COMH93D67098 Combi-Blocks, Inc.

N-(4-Fluorobenzyl)-4-methoxyaniline

  • MDL number MFCD03210757
  • Cas Number : 356531-43-6
  • Molecular Weight : 231.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-6319

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-6319
Availability details for
Quantity
  • Availability
  • Add to Cart