COMH93D670A2 Combi-Blocks, Inc.

2-Methyl-4-(trifluoromethoxy)-dl-phenylalanine

  • MDL number MFCD22201062
  • Cas Number : 1706428-65-0
  • Molecular Weight : 263.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-6359

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-6359
Availability details for
Quantity
  • Availability
  • Add to Cart