COMH93D670C7 Combi-Blocks, Inc.

2-[5-(4-Pyridyl)-1h-1,2,4-triazol-3-yl]pyridine

  • MDL number MFCD00101970
  • Cas Number : 36770-50-0
  • Molecular Weight : 223.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-6601

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-6601
Availability details for
Quantity
  • Availability
  • Add to Cart