COMH93D670EA Combi-Blocks, Inc.

5-Fluoro-2-(trifluoromethoxy)benzonitrile

  • MDL number MFCD11519332
  • Cas Number : 1092460-82-6
  • Molecular Weight : 205.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-6777

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-6777
Availability details for
Quantity
  • Availability
  • Add to Cart