COMH93D670EC Combi-Blocks, Inc.

N,N-Diisopropyl-3-methylbenzamide

  • MDL number MFCD00587389
  • Cas Number : 5448-36-2
  • Molecular Weight : 219.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-6796

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-6796
Availability details for
Quantity
  • Availability
  • Add to Cart