COMH93D670FD Combi-Blocks, Inc.

7-Fluoro-6-nitro-2h-1,4-benzoxazin-3(4h)-one

  • MDL number MFCD22056242
  • Cas Number : 103361-67-7
  • Molecular Weight : 212.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-6912

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-6912
Availability details for
Quantity
  • Availability
  • Add to Cart