COMH93D6711A Combi-Blocks, Inc.

1,4-Bis(4-pyridylmethyl)benzene

  • MDL number MFCD23379844
  • Cas Number : 52469-20-2
  • Molecular Weight : 260.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-7105

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-7105
Availability details for
Quantity
  • Availability
  • Add to Cart