COMH93D6712B Combi-Blocks, Inc.

(S)-(-)-1-(4-Bromophenyl)ethyl isocyanate

  • MDL number MFCD05664122
  • Cas Number : 149552-52-3
  • Molecular Weight : 226.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-7231

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-7231
Availability details for
Quantity
  • Availability
  • Add to Cart