COMH93D67156 Combi-Blocks, Inc.

N-Ethyl-2-(4-piperidinyloxy)acetamide

  • MDL number MFCD08061052
  • Cas Number : 521074-81-7
  • Molecular Weight : 186.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-7515

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-7515
Availability details for
Quantity
  • Availability
  • Add to Cart