COMH93D67178 Combi-Blocks, Inc.

2-Bromo-4-(2-furyl)thiazole

  • MDL number MFCD11220134
  • Cas Number : 917566-86-0
  • Molecular Weight : 230.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-bromo-4-(2-furyl)-1,3-thiazole
Inchi Code 1S/C7H4BrNOS/c8-7-9-5(4-11-7)6-2-1-3-10-6/h1-4H
Inchi Key PUFWLSRDFTVPMS-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-7707

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-7707
Availability details for
Quantity
  • Availability
  • Add to Cart