COMH93D67184 Combi-Blocks, Inc.

2-(4-Piperidinyloxy)-n-propylacetamide

  • MDL number MFCD08064265
  • Cas Number : 902836-22-0
  • Molecular Weight : 200.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-7778

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-7778
Availability details for
Quantity
  • Availability
  • Add to Cart