COMH93D671A8 Combi-Blocks, Inc.

4-Methylbenzanilide

  • MDL number MFCD00017213
  • Cas Number : 6833-18-7
  • Molecular Weight : 211.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-methyl-N-phenylbenzamide
Inchi Code 1S/C14H13NO/c1-11-7-9-12(10-8-11)14(16)15-13-5-3-2-4-6-13/h2-10H,1H3,(H,15,16)
Inchi Key PQEOPHYIUYAVDQ-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-8039

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-8039
Availability details for
Quantity
  • Availability
  • Add to Cart