COMH93D671C0 Combi-Blocks, Inc.

3-(3-Ethyl-1,2,4-oxadiazol-5-yl)piperidine

  • MDL number MFCD08061044
  • Cas Number : 139269-06-0
  • Molecular Weight : 181.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(3-ethyl-1,2,4-oxadiazol-5-yl)piperidine
Inchi Code 1S/C9H15N3O/c1-2-8-11-9(13-12-8)7-4-3-5-10-6-7/h7,10H,2-6H2,1H3
Inchi Key XXIODYOSZOJCLL-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-8215

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-8215
Availability details for
Quantity
  • Availability
  • Add to Cart