COMH93D671DD Combi-Blocks, Inc.

2-Methoxy-4,6-bis(trifluoromethyl)aniline

  • MDL number MFCD11519320
  • Cas Number : 1092460-74-6
  • Molecular Weight : 259.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-8445

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-8445
Availability details for
Quantity
  • Availability
  • Add to Cart