COMH93D671DE Combi-Blocks, Inc.

5-Methoxy-1-methyl-4-nitroindole-3-carboxaldehyde

  • MDL number MFCD03093098
  • Cas Number : 191846-76-1
  • Molecular Weight : 234.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-8451

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-8451
Availability details for
Quantity
  • Availability
  • Add to Cart