COMH93D671E9 Combi-Blocks, Inc.

2-(N-Boc-methylamino)-5-iodo-3-methylpyridine

  • MDL number MFCD09037457
  • Cas Number : 942206-08-8
  • Molecular Weight : 348.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl 5-iodo-3-methyl-2-pyridinyl(methyl)carbamate
Inchi Code 1S/C12H17IN2O2/c1-8-6-9(13)7-14-10(8)15(5)11(16)17-12(2,3)4/h6-7H,1-5H3
Inchi Key HWHYESISLPNYCT-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-8499

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-8499
Availability details for
Quantity
  • Availability
  • Add to Cart