COMH93D671F8 Combi-Blocks, Inc.

4-Chloro-n-(4-fluorobenzyl)-2-methylaniline

  • MDL number MFCD11119360
  • Cas Number : 1036628-11-1
  • Molecular Weight : 249.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-chloro-N-(4-fluorobenzyl)-2-methylaniline
Inchi Code 1S/C14H13ClFN/c1-10-8-12(15)4-7-14(10)17-9-11-2-5-13(16)6-3-11/h2-8,17H,9H2,1H3
Inchi Key CYUBYUVQPNJDHU-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-8570

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-8570
Availability details for
Quantity
  • Availability
  • Add to Cart