COMH93D671FD Combi-Blocks, Inc.

N-(4,6-Dimethyl-2-pyridyl)thiourea

  • MDL number MFCD15525456
  • Cas Number : 49600-35-3
  • Molecular Weight : 181.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(4,6-dimethylpyridin-2-yl)thiourea
Inchi Code 1S/C8H11N3S/c1-5-3-6(2)10-7(4-5)11-8(9)12/h3-4H,1-2H3,(H3,9,10,11,12)
Inchi Key ZXQTZSMIBLLDGW-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-8612

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-8612
Availability details for
Quantity
  • Availability
  • Add to Cart