COMH93D67211 Combi-Blocks, Inc.

3-Methoxy-2-(trifluoromethyl)benzylamine

  • MDL number MFCD18398592
  • Cas Number : 1261647-71-5
  • Molecular Weight : 205.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-8728

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-8728
Availability details for
Quantity
  • Availability
  • Add to Cart