COMH93D67216 Combi-Blocks, Inc.

1,1,3,5,5-Pentaphenyl-1,3,5-triphosphapentane trioxide

  • MDL number MFCD00482218
  • Cas Number : 21851-89-8
  • Molecular Weight : 554.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-8767

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-8767
Availability details for
Quantity
  • Availability
  • Add to Cart