COMH93D6726E Combi-Blocks, Inc.

2-Chloro-5-(pentafluorosulfur)benzoic acid

  • MDL number MFCD22988931
  • Cas Number : 1431329-70-2
  • Molecular Weight : 282.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-9368

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-9368
Availability details for
Quantity
  • Availability
  • Add to Cart