COMH93D6733A Combi-Blocks, Inc.

2',5-Dichloro-2-hydroxybenzophenone

  • MDL number MFCD02682994
  • Cas Number : 61785-35-1
  • Molecular Weight : 267.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (5-chloro-2-hydroxyphenyl)(2-chlorophenyl)methanone
Inchi Code 1S/C13H8Cl2O2/c14-8-5-6-12(16)10(7-8)13(17)9-3-1-2-4-11(9)15/h1-7,16H
Inchi Key OSXVZDOVYCTYCW-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-1552

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-1552
Availability details for
Quantity
  • Availability
  • Add to Cart