COMH93D67345 Combi-Blocks, Inc.

1,2-Epoxycyclododecane

  • MDL number MFCD00005116
  • Cas Number : 286-99-7
  • Molecular Weight : 182.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 13-oxabicyclo[10.1.0]tridecane
Inchi Code 1S/C12H22O/c1-2-4-6-8-10-12-11(13-12)9-7-5-3-1/h11-12H,1-10H2
Inchi Key VLJLXEKIAALSJE-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-1739

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-1739
Availability details for
Quantity
  • Availability
  • Add to Cart