COMH93D67487 Combi-Blocks, Inc.

Cis-3-methyl-2-pentene

  • MDL number MFCD00066518
  • Cas Number : 922-62-3
  • Molecular Weight : 84.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-6549

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-6549
Availability details for
Quantity
  • Availability
  • Add to Cart