COMH93D67538 Combi-Blocks, Inc.

Dl-2-methylglutamic acid hemihydrate

  • MDL number MFCD00149281
  • Cas Number : 71-90-9
  • Molecular Weight : 179.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-methylglutamic acid hydrate
Inchi Code 1S/C6H11NO4.H2O/c1-6(7,5(10)11)3-2-4(8)9;/h2-3,7H2,1H3,(H,8,9)(H,10,11);1H2
Inchi Key UAHXIVOASZLFEG-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-9163

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-9163
Availability details for
Quantity
  • Availability
  • Add to Cart