COMH93D67578 Combi-Blocks, Inc.

4-Bromo-3,3,4,4-tetrafluorobutyl 1h,1h,2h,2h-perfluorohexyl carbonate

  • MDL number MFCD28334466
  • Cas Number : 1980035-10-6
  • Molecular Weight : 515.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-bromo-3,3,4,4-tetrafluorobutyl (3,3,4,4,5,5,6,6,6-nonafluorohexyl) carbonate
Inchi Code 1S/C11H8BrF13O3/c12-10(21,22)7(15,16)2-4-28-5(26)27-3-1-6(13,14)8(17,18)9(19,20)11(23,24)25/h1-4H2
Inchi Key NBMCUJSQJZMTKT-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QI-0028

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QI-0028
Availability details for
Quantity
  • Availability
  • Add to Cart