COMH93D67585 Combi-Blocks, Inc.

4-(2-Aminoethylthio)-8-fluoro-2-(trifluoromethyl)quinoline

  • MDL number MFCD04038213
  • Cas Number : 541539-78-0
  • Molecular Weight : 290.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-{[8-fluoro-2-(trifluoromethyl)-4-quinolinyl]sulfanyl}ethanamine
Inchi Code 1S/C12H10F4N2S/c13-8-3-1-2-7-9(19-5-4-17)6-10(12(14,15)16)18-11(7)8/h1-3,6H,4-5,17H2
Inchi Key ORBZAAFUCWIVQM-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QI-0090

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QI-0090
Availability details for
Quantity
  • Availability
  • Add to Cart