COMH93D675C9 Combi-Blocks, Inc.

(3-(tert-Butyl)phenyl)methanamine

  • MDL number MFCD09260785
  • Cas Number : 140401-55-4
  • Molecular Weight : 163.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3-tert-butylphenyl)methanamine
Inchi Code 1S/C11H17N/c1-11(2,3)10-6-4-5-9(7-10)8-12/h4-7H,8,12H2,1-3H3
Inchi Key JPETYCDJGPTICO-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QI-0344

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QI-0344
Availability details for
Quantity
  • Availability
  • Add to Cart