COMH93D675FA Combi-Blocks, Inc.

2,2-Difluoroethyl 2,2,3,3-tetrafluoropropyl carbonate

  • MDL number MFCD28334294
  • Cas Number : 1980045-84-8
  • Molecular Weight : 240.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2,2-difluoroethyl (2,2,3,3-tetrafluoropropyl) carbonate
Inchi Code 1S/C6H6F6O3/c7-3(8)1-14-5(13)15-2-6(11,12)4(9)10/h3-4H,1-2H2
Inchi Key DCJNEQBBLMAAMI-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QI-0519

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QI-0519
Availability details for
Quantity
  • Availability
  • Add to Cart