COMH93D675FF Combi-Blocks, Inc.

2-Fluoro-4-(phenylsulfonyl)phenylhydrazine

  • MDL number MFCD09800751
  • Cas Number : 1000339-96-7
  • Molecular Weight : 266.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QI-0530

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QI-0530
Availability details for
Quantity
  • Availability
  • Add to Cart