COMH93D6760A Combi-Blocks, Inc.

Methyl 3-amino-5-(hydroxymethyl)benzoate

  • MDL number MFCD11036276
  • Cas Number : 220286-47-5
  • Molecular Weight : 181.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 3-amino-5-(hydroxymethyl)benzoate
Inchi Code 1S/C9H11NO3/c1-13-9(12)7-2-6(5-11)3-8(10)4-7/h2-4,11H,5,10H2,1H3
Inchi Key MOYDZZVEYJZZGZ-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QI-0590

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QI-0590
Availability details for
Quantity
  • Availability
  • Add to Cart