COMH93D67645 Combi-Blocks, Inc.

2,3,5-Trifluoro-4-hydrazinobenzohydrazide

  • MDL number MFCD27995621
  • Cas Number : 1823962-17-9
  • Molecular Weight : 220.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QI-0832

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QI-0832
Availability details for
Quantity
  • Availability
  • Add to Cart