COMH93D6765F Combi-Blocks, Inc.

Methyl 3-acetylbicyclo[1.1.1]pentane-1-carboxylate

  • MDL number MFCD24696091
  • Cas Number : 131515-42-9
  • Molecular Weight : 168.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QI-0985

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QI-0985
Availability details for
Quantity
  • Availability
  • Add to Cart