COMH93D67670 Combi-Blocks, Inc.

3-Butoxy-2,6-difluorobenzyl alcohol

  • MDL number MFCD27986841
  • Cas Number : 1706435-02-0
  • Molecular Weight : 216.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QI-1072

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QI-1072
Availability details for
Quantity
  • Availability
  • Add to Cart