COMH93D676A0 Combi-Blocks, Inc.

Methyl 2-(4-nitrophenyl)-2-(4-bromophenyl)acetate

  • MDL number MFCD28154747
  • Cas Number : 1980044-76-5
  • Molecular Weight : 350.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QI-1337

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QI-1337
Availability details for
Quantity
  • Availability
  • Add to Cart