COMH93D677F7 Combi-Blocks, Inc.

N'-[1-Amino-2-(4-methoxyphenyl)ethylidene]hydrazinecarboxylic acid tert-butyl ester

  • MDL number MFCD10568184
  • Cas Number : 1053655-79-0
  • Molecular Weight : 279.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QI-2837

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QI-2837
Availability details for
Quantity
  • Availability
  • Add to Cart