COMH93D678D4 Combi-Blocks, Inc.

Chaetocin

  • MDL number MFCD00133163
  • Cas Number : 28097-03-2
  • Molecular Weight : 696.85

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QI-5452

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QI-5452
Availability details for
Quantity
  • Availability
  • Add to Cart