COMH93D6791E Combi-Blocks, Inc.

2,7-Dimethoxy-9h-carbazole

  • MDL number MFCD28154716
  • Cas Number : 61822-18-2
  • Molecular Weight : 227.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QI-7061

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QI-7061
Availability details for
Quantity
  • Availability
  • Add to Cart