COMH93D6794E Combi-Blocks, Inc.

Perfluoro(2-methyl-3-pentanone)

  • MDL number MFCD04038343
  • Cas Number : 756-13-8
  • Molecular Weight : 316.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QI-8073

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QI-8073
Availability details for
Quantity
  • Availability
  • Add to Cart