COMH93D6799B Combi-Blocks, Inc.

Ethyl 4-(diethoxyphosphoryl)-3-methylcrotonate

  • MDL number MFCD08445507
  • Cas Number : 41891-54-7
  • Molecular Weight : 264.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-0434

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-0434
Availability details for
Quantity
  • Availability
  • Add to Cart