COMH93D679A6 Combi-Blocks, Inc.

1-(Furo[3,2-c]pyridin-4-yl)homopiperazine

  • MDL number MFCD06658987
  • Cas Number : 845885-87-2
  • Molecular Weight : 217.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-(1,4-diazepan-1-yl)furo[3,2-c]pyridine
Inchi Code 1S/C12H15N3O/c1-4-13-6-8-15(7-1)12-10-3-9-16-11(10)2-5-14-12/h2-3,5,9,13H,1,4,6-8H2
Inchi Key JGVLLHBTSVQMHS-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-0967

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-0967
Availability details for
Quantity
  • Availability
  • Add to Cart