COMH93D67A1B Combi-Blocks, Inc.

(3-Methoxythien-2-yl)methylamine

  • MDL number MFCD11841071
  • Cas Number : 946409-37-6
  • Molecular Weight : 143.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3-methoxy-2-thienyl)methanamine
Inchi Code 1S/C6H9NOS/c1-8-5-2-3-9-6(5)4-7/h2-3H,4,7H2,1H3
Inchi Key GDDVAKBOBBYCLK-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-4899

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-4899
Availability details for
Quantity
  • Availability
  • Add to Cart