COMH93D67AD1 Combi-Blocks, Inc.

6-(Chloromethyl)quinoxaline

  • MDL number MFCD15144408
  • Cas Number : 477776-17-3
  • Molecular Weight : 178.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QK-2291

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QK-2291
Availability details for
Quantity
  • Availability
  • Add to Cart