COMH93D67D00 Combi-Blocks, Inc.

1-Methyl-1-propylpiperidinium bromide

  • MDL number MFCD17014743
  • Cas Number : 88840-42-0
  • Molecular Weight : 222.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QN-4809

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QN-4809
Availability details for
Quantity
  • Availability
  • Add to Cart