COMH93D67D25 Combi-Blocks, Inc.

1,2,3,4,5,6-Hexa-o-[11-[4-(4-hexylphenylazo)phenoxy]undecanoyl]-d-mannitol

  • MDL number MFCD28100832
  • Cas Number : 1093077-77-0
  • Molecular Weight : 2874.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QN-5520

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QN-5520
Availability details for
Quantity
  • Availability
  • Add to Cart