COMH93D67D6C Combi-Blocks, Inc.

Allyl nonanoate

  • MDL number MFCD00038344
  • Cas Number : 7493-72-3
  • Molecular Weight : 198.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QN-6694

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QN-6694
Availability details for
Quantity
  • Availability
  • Add to Cart