COMH93D67D8F Combi-Blocks, Inc.

1,2-Bis(4-methoxyphenyl)-2-oxoethyl cyclohexylcarbamate

  • MDL number MFCD29472516
  • Cas Number : 196599-80-1
  • Molecular Weight : 397.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QN-7148

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QN-7148
Availability details for
Quantity
  • Availability
  • Add to Cart