COMH93D67D9D Combi-Blocks, Inc.

Isobutyl n-octanoate

  • MDL number MFCD00059444
  • Cas Number : 5461-06-3
  • Molecular Weight : 200.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QN-7391

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QN-7391
Availability details for
Quantity
  • Availability
  • Add to Cart