COMH93D67DC0 Combi-Blocks, Inc.

Tris(1,10-phenanthroline)ruthenium(ii) bis(hexafluorophosphate)

  • MDL number MFCD28369263
  • Cas Number : 60804-75-3
  • Molecular Weight : 931.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QN-7858

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QN-7858
Availability details for
Quantity
  • Availability
  • Add to Cart