COMH93D67DC1 Combi-Blocks, Inc.

4-(2,2-Diphenylethenyl)-n,n-di(p-tolyl)aniline

  • MDL number MFCD09832972
  • Cas Number : 89114-91-0
  • Molecular Weight : 451.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QN-7863

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QN-7863
Availability details for
Quantity
  • Availability
  • Add to Cart